Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ile-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060147.6 | NP_492548.1 | 2 - 3 | 156 | 68 | |
GenBank mRNA | DQ384615.1 | - | 2 - 3 | 154 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |