Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tba-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001264283.3 | NP_001251212.1 | 2 - 3 | 844 | 77 | |
NM_001383674.1 | 3 - 4 | 930 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Endogenous Controls Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |