Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mps-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058377.4 | NP_490778.1 | 1 - 2 | 54 | 89 | |
GenBank mRNA | AY255668.1 | - | 1 - 2 | 41 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |