Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306500.3 | NP_001293429.1 | 5 - 6 | 702 | 95 | |
NM_058459.6 | NP_490860.1 | 6 - 7 | 959 | 95 | ||
GenBank mRNA | AJ131742.1 | - | 6 - 7 | 936 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |