Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: icp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306434.3 | NP_001293363.1 | 3 - 4 | 1589 | 84 | |
NM_058555.9 | NP_490956.1 | 4 - 5 | 1746 | 84 | ||
GenBank mRNA | AF300704.1 | - | 4 - 5 | 1748 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |