Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fog-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381585.1 | NP_001367957.1 | 1 - 2 | 80 | 63 | |
NM_001381586.2 | NP_001367956.1 | 1 - 2 | 86 | 63 | ||
GenBank mRNA | AJ297846.1 | - | 1 - 2 | 102 | 63 | |
AJ297847.1 | - | 1 - 2 | 102 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |