Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fog-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381585.1 | NP_001367957.1 | 5 - 6 | 745 | 74 | |
NM_001381586.2 | NP_001367956.1 | 5 - 6 | 751 | 74 | ||
GenBank mRNA | AJ297846.1 | - | 5 - 6 | 767 | 74 | |
AJ297847.1 | - | 5 - 6 | 767 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |