Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sulp-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373791.2 | NP_001360427.1 | 4 - 5 | 921 | 67 | |
NM_058737.5 | NP_491138.2 | 4 - 5 | 904 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |