Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026429.3 | NP_001021600.1 | 16 - 17 | 3690 | 103 | |
NM_001262699.2 | NP_001249628.1 | 19 - 20 | 3584 | 103 | ||
NM_001382937.1 | 15 - 16 | 3481 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |