Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phat-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059108.6 | NP_491509.1 | 3 - 4 | 648 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |