Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059140.6 | NP_491541.1 | 6 - 7 | 1049 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |