Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: erm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059158.7 | NP_491559.1 | 7 - 8 | 1076 | 63 | |
NM_059159.4 | NP_491560.1 | 7 - 8 | 1042 | 63 | ||
GenBank mRNA | AB107268.1 | - | 7 - 8 | 1098 | 63 | |
AB107269.1 | - | 7 - 8 | 1064 | 63 | ||
AY643538.1 | - | 7 - 8 | 1066 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |