Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026159.6 | NP_001021330.1 | 6 - 7 | 1320 | 89 | |
NM_171391.7 | NP_741456.1 | 5 - 6 | 1385 | 89 | ||
NR_131580.1 | 5 - 6 | 1266 | 89 | |||
GenBank mRNA | AF273776.1 | - | 5 - 6 | 1375 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |