Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307091.3 | NP_001294020.1 | 3 - 4 | 199 | 71 | |
NM_171418.7 | NP_741488.1 | 3 - 4 | 197 | 71 | ||
NM_171918.7 | NP_741489.1 | 3 - 4 | 199 | 71 | ||
GenBank mRNA | X68070.1 | - | 206 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |