Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-35C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171550.5 | NP_741638.1 | 3 - 4 | 485 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |