Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171587.6 | NP_741677.1 | 1 | 1117 | 69 | |
GenBank mRNA | AF208526.1 | - | 1 | 1141 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Both primers and probe map within a single exon |