Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171659.6 | NP_741758.1 | 7 - 8 | 1563 | 64 | |
GenBank mRNA | U16367.1 | - | 7 - 8 | 1563 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |