Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coel-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171665.6 | NP_741764.1 | 4 - 5 | 612 | 84 | |
NM_171947.7 | NP_741765.1 | 5 - 6 | 758 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |