Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: paf-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171668.9 | NP_741768.1 | 3 - 4 | 461 | 131 | |
NM_171669.6 | NP_741769.1 | 2 - 3 | 278 | 131 | ||
GenBank mRNA | AF386745.1 | - | 2 - 3 | 277 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |