Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027202.7 | NP_001022373.1 | 6 - 7 | 722 | 89 | |
NM_001027203.5 | NP_001022374.1 | 6 - 7 | 724 | 89 | ||
GenBank mRNA | D84668.1 | - | 6 - 7 | 722 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |