Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379816.2 | NP_001367451.1 | 7 - 8 | 1301 | 74 | |
NM_001379817.1 | NP_001367450.1 | 6 - 7 | 1637 | 74 | ||
NM_066339.6 | NP_498740.2 | 6 - 7 | 1321 | 74 | ||
NM_066340.6 | NP_498741.3 | 8 - 9 | 1534 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |