Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttyh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383646.1 | 5 - 6 | 985 | 115 | ||
NM_077502.7 | NP_509903.1 | 4 - 5 | 852 | 115 | ||
NM_171973.9 | NP_741904.1 | 4 - 5 | 854 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |