Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exo-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026413.6 | NP_001021584.1 | 2 - 3 | 597 | 98 | |
GenBank mRNA | AF034258.1 | - | 2 - 3 | 591 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |