Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061001.1 | NP_493402.1 | 6 - 7 | 1205 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |