Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383767.1 | 7 - 8 | 917 | 87 | ||
NM_061181.7 | NP_493582.3 | 7 - 8 | 910 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |