Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dog-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061217.6 | NP_493618.1 | 13 - 14 | 2767 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |