Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313675.3 | NP_001300604.1 | 1 | 195 | 81 | |
NM_001377684.3 | NP_001364775.1 | 31 - 32 | 7576 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |