Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxc-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061871.6 | NP_494272.1 | 4 - 5 | 2151 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |