Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fut-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061971.5 | NP_494372.2 | 3 - 4 | 385 | 121 | |
GenBank mRNA | AJ505020.1 | - | 3 - 4 | 326 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |