Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062273.1 | NP_494674.1 | 7 - 8 | 1419 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |