Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhgf-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381382.1 | NP_001368191.1 | 7 - 8 | 1373 | 98 | |
NM_001381384.1 | NP_001367992.1 | 7 - 8 | 1104 | 98 | ||
NM_001383815.1 | 7 - 8 | 1107 | 98 | |||
GenBank mRNA | JQ763434.1 | - | 6 - 7 | 947 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |