Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027161.3 | NP_001022332.1 | 8 - 9 | 1459 | 97 | |
NM_001377823.1 | NP_001364642.1 | 8 - 9 | 1432 | 97 | ||
NM_001377824.1 | NP_001364641.1 | 8 - 9 | 1408 | 97 | ||
NM_001377825.1 | NP_001364552.1 | 8 - 9 | 1381 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |