Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sfxn-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026851.6 | NP_001022022.1 | 4 - 5 | 913 | 63 | |
NM_001322747.3 | NP_001309542.1 | 3 - 4 | 618 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |