Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027088.7 | NP_001022259.1 | 6 - 7 | 1697 | 75 | |
NM_001027089.7 | NP_001022260.1 | 5 - 6 | 1565 | 75 | ||
NM_001373755.2 | NP_001359723.1 | 5 - 6 | 1584 | 75 | ||
NM_001381521.1 | NP_001368728.1 | 5 - 6 | 1558 | 75 | ||
NM_001383902.1 | 6 - 7 | 1658 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |