Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063479.5 | NP_495880.1 | 1 - 2 | 90 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |