Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nuo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063975.5 | NP_496376.1 | 4 - 5 | 1026 | 66 |
Gene Symbol: C09H10.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383946.1 | 4 - 5 | 1013 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |