Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clh-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026729.4 | NP_001021900.1 | 4 - 5 | 484 | 113 | |
NM_001377875.3 | NP_001364585.1 | 4 - 5 | 562 | 113 | ||
NM_001383953.1 | 3 - 4 | 372 | 113 | |||
GenBank mRNA | AF173171.1 | - | 3 - 4 | 394 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |