Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clh-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026729.4 | NP_001021900.1 | 5 - 6 | 623 | 65 | |
NM_001377875.3 | NP_001364585.1 | 5 - 6 | 701 | 65 | ||
NM_001383953.1 | 4 - 5 | 511 | 65 | |||
GenBank mRNA | AF173171.1 | - | 4 - 5 | 533 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |