Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027267.5 | NP_001022438.1 | 1 - 2 | 98 | 94 | |
NM_001373808.1 | NP_001359643.1 | 1 - 2 | 93 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |