Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gsy-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064335.6 | NP_496736.1 | 2 - 3 | 194 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |