Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064540.6 | NP_496941.1 | 2 - 3 | 364 | 79 | |
GenBank mRNA | AY438643.1 | - | 2 - 3 | 364 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |