Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mog-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064626.5 | NP_497027.1 | 8 - 9 | 2903 | 94 | |
GenBank mRNA | AF286900.1 | - | 8 - 9 | 2919 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |