Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mat-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064802.6 | NP_497203.1 | 4 - 5 | 858 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |