Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065850.3 | NP_498251.1 | 1 - 2 | 136 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |