Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cup-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027550.4 | NP_001022721.2 | 2 - 3 | 523 | 112 | |
NM_001027551.5 | NP_001022722.2 | 1 - 2 | 463 | 112 | ||
NM_001268016.4 | NP_001254945.1 | 1 - 2 | 463 | 112 | ||
NM_001382919.1 | 1 - 2 | 472 | 112 | |||
NM_066263.6 | NP_498664.3 | 1 - 2 | 458 | 112 | ||
GenBank mRNA | AF482952.1 | - | 1 - 2 | 457 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |