Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sod-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268073.3 | NP_001255002.1 | 3 - 4 | 237 | 133 | |
NM_001268074.4 | NP_001255003.1 | 3 - 4 | 299 | 133 | ||
GenBank mRNA | AB003924.1 | - | 2 - 3 | 164 | 133 | |
AB190513.1 | - | 2 - 3 | 164 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |