Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hlh-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379865.1 | NP_001367058.1 | 3 - 4 | 267 | 70 | |
NM_066728.7 | NP_499129.2 | 3 - 4 | 301 | 70 | ||
NM_066729.9 | NP_499130.2 | 3 - 4 | 300 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |