Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306672.3 | NP_001293601.1 | 3 - 4 | 746 | 82 | |
NM_066775.6 | NP_499176.1 | 5 - 6 | 1289 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |