Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zyg-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067170.7 | NP_499571.2 | 2 - 3 | 275 | 93 | |
GenBank mRNA | AJ319870.1 | - | 1 - 2 | 190 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |