Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plk-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068795.7 | NP_501196.1 | 6 - 7 | 1573 | 63 | |
GenBank mRNA | AF059024.1 | - | 6 - 7 | 1573 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |